Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691155.3 | XP_005691212.2 | 3 - 4 | 436 | 86 | |
| XM_005691156.3 | XP_005691213.2 | 3 - 4 | 436 | 86 | ||
| GenBank mRNA | JO420002.2 | - | 6 - 7 | 1516 | 86 | |
More Information
Set Membership: |
|

FGG
Genomic Map