Assay Details
| Gene Symbol: NUTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692159.3 | XP_005692216.1 | 4 - 5 | 731 | 79 | |
| XM_005692160.3 | XP_005692217.1 | 4 - 5 | 418 | 79 | ||
More Information
Set Membership: |
|

NUTF2
Genomic Map