Assay Details
| Gene Symbol: RPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692683.3 | XP_005692740.1 | 3 - 4 | 305 | 52 | |
| GenBank mRNA | JO375941.2 | - | 4 - 5 | 412 | 52 | |
| JO580688.1 | - | 4 - 5 | 473 | 52 | ||
More Information
Set Membership: |
|

RPL18
Genomic Map