Assay Details
| Gene Symbol: SPAG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693257.3 | XP_005693314.2 | 22 - 23 | 3812 | 61 | |
| XM_013971868.2 | XP_013827322.2 | 22 - 23 | 3424 | 61 | ||
| XM_018064309.1 | XP_017919798.1 | 22 - 23 | 3464 | 61 | ||
| GenBank mRNA | JO600017.2 | - | 2 - 3 | 240 | 61 | |
| JT703898.1 | - | 3 - 4 | 348 | 61 | ||
More Information
Set Membership: |
|

SPAG5
Genomic Map