Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065015.1 | XP_017920504.1 | 2 - 3 | 387 | 63 | |
| XM_018065016.1 | XP_017920505.1 | 2 - 3 | 393 | 63 | ||
| GenBank mRNA | JO429919.2 | - | 2 - 3 | 1763 | 63 | |
More Information
Set Membership: |
|

ORMDL3
Genomic Map