Assay Details
| Gene Symbol: VPS52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696219.3 | XP_005696276.1 | 10 - 11 | 1253 | 57 | |
| XM_013973825.2 | XP_013829279.1 | 10 - 11 | 965 | 57 | ||
| XM_013973826.2 | XP_013829280.1 | 10 - 11 | 1211 | 57 | ||
| GenBank mRNA | JO634262.2 | - | 10 - 11 | 1031 | 57 | |
More Information
Set Membership: |
|

VPS52
Genomic Map