Assay Details
| Gene Symbol: HNRNPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699108.3 | XP_005699165.1 | 5 - 6 | 766 | 76 | |
| XM_005699109.3 | XP_005699166.1 | 5 - 6 | 747 | 76 | ||
| XM_005699110.3 | XP_005699167.1 | 5 - 6 | 721 | 76 | ||
| XM_005699111.3 | XP_005699168.1 | 5 - 6 | 702 | 76 | ||
| XM_018042223.1 | XP_017897712.1 | 5 - 6 | 738 | 76 | ||
| XM_018042225.1 | XP_017897714.1 | 4 - 5 | 962 | 76 | ||
| XM_018042226.1 | XP_017897715.1 | 4 - 5 | 608 | 76 | ||
| XM_018042227.1 | XP_017897716.1 | 4 - 5 | 587 | 76 | ||
| XM_018042228.1 | XP_017897717.1 | 4 - 5 | 563 | 76 | ||
| GenBank mRNA | JO580000.1 | - | 5 - 6 | 797 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018042227.1, XM_018042228.1 transcripts.
Set Membership: |
|

HNRNPH3
Genomic Map