Assay Details
| Gene Symbol: PPP1R3F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700737.3 | XP_005700794.3 | 3 - 4 | 4203 | 79 | |
| XM_013976470.2 | XP_013831924.2 | 3 - 4 | 4200 | 79 | ||
| XM_018045138.1 | XP_017900627.1 | 3 - 4 | 4128 | 79 | ||
| GenBank mRNA | JO639149.2 | - | 1 - 2 | 1758 | 79 | |
| JT705173.1 | - | 8 - 9 | 1408 | 79 | ||
More Information
Set Membership: |
|

PPP1R3F
Genomic Map