Assay Details
| Gene Symbol: REPIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046776.1 | XP_017902265.1 | 2 - 3 | 263 | 97 | |
| XM_018046778.1 | XP_017902267.1 | 2 - 3 | 334 | 97 | ||
More Information
Set Membership: |
|

REPIN1
Genomic Map