Assay Details
| Gene Symbol: ZNF775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046774.1 | XP_017902263.1 | 2 - 3 | 895 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018046774.1 transcripts.
Set Membership: |
|

ZNF775
Genomic Map