Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048203.1 | XP_017903692.1 | 18 - 19 | 1881 | 67 | |
| XM_018048204.1 | XP_017903693.1 | 15 - 16 | 1504 | 67 | ||
| GenBank mRNA | JT666958.1 | - | 18 - 19 | 1866 | 67 | |
| JT689199.1 | - | 18 - 19 | 1852 | 67 | ||
More Information
Set Membership: |
|

WASHC4
Genomic Map