Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048203.1 | XP_017903692.1 | 23 - 24 | 2501 | 159 | |
XM_018048204.1 | XP_017903693.1 | 20 - 21 | 2124 | 159 | ||
GenBank mRNA | JT666958.1 | - | 23 - 24 | 2480 | 159 | |
JT689199.1 | - | 23 - 24 | 2455 | 159 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
