Assay Details
| Gene Symbol: WASHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048203.1 | XP_017903692.1 | 9 - 10 | 752 | 100 | |
| XM_018048204.1 | XP_017903693.1 | 6 - 7 | 375 | 100 | ||
| GenBank mRNA | JT666958.1 | - | 9 - 10 | 736 | 100 | |
More Information
Set Membership: |
|

WASHC4
Genomic Map