Assay Details
| Gene Symbol: CCDC187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056087.1 | XP_017911576.1 | 17 - 18 | 4159 | 99 | |
| XM_018056088.1 | XP_017911577.1 | 17 - 18 | 4159 | 99 | ||
| XM_018056090.1 | XP_017911579.1 | 16 - 17 | 4063 | 99 | ||
| XM_018056091.1 | XP_017911580.1 | 19 - 20 | 4060 | 99 | ||
| XM_018056092.1 | XP_017911581.1 | 18 - 19 | 3946 | 99 | ||
| GenBank mRNA | KA352270.1 | - | 2 - 3 | 318 | 99 | |
More Information
Set Membership: |
|

CCDC187
Genomic Map