Assay Details
| Gene Symbol: CCDC187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056087.1 | XP_017911576.1 | 19 - 20 | 4414 | 103 | |
| XM_018056089.1 | XP_017911578.1 | 19 - 20 | 4405 | 103 | ||
| XM_018056090.1 | XP_017911579.1 | 18 - 19 | 4318 | 103 | ||
| XM_018056091.1 | XP_017911580.1 | 21 - 22 | 4315 | 103 | ||
| XM_018056092.1 | XP_017911581.1 | 20 - 21 | 4201 | 103 | ||
| GenBank mRNA | KA352270.1 | - | 4 - 5 | 573 | 103 | |
More Information
Set Membership: |
|

CCDC187
Genomic Map