Assay Details
| Gene Symbol: PTPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058021.1 | XP_017913510.1 | 3 - 4 | 418 | 58 | |
| XM_018058022.1 | XP_017913511.1 | 3 - 4 | 267 | 58 | ||
| GenBank mRNA | JO410306.2 | - | 3 - 4 | 458 | 58 | |
| KA334315.1 | - | 6 - 7 | 1301 | 58 | ||
More Information
Set Membership: |
|

PTPN1
Genomic Map