Assay Details
| Gene Symbol: C15H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059831.1 | XP_017915320.1 | 7 - 8 | 823 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15H11orf65| Gene Symbol: C15H11orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059831.1 | XP_017915320.1 | 7 - 8 | 823 | 91 | |
Set Membership: |
|