Assay Details
| Gene Symbol: MDK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059158.1 | XP_017914647.1 | 4 - 5 | 551 | 59 | |
| XM_018059159.1 | XP_017914648.1 | 4 - 5 | 528 | 59 | ||
| GenBank mRNA | JO447329.2 | - | 1 - 2 | 340 | 59 | |
More Information
Set Membership: |
|

MDK
Genomic Map