Assay Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013970610.2 | XP_013826064.2 | 6 - 7 | 962 | 59 | |
| XM_018061223.1 | XP_017916712.1 | 7 - 8 | 1124 | 59 | ||
| GenBank mRNA | JT710081.1 | - | 7 - 8 | 1078 | 59 | |
More Information
Set Membership: |
|

WSB2
Genomic Map