Assay Details
| Gene Symbol: UPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044912.1 | XP_017900401.1 | 968 | 60 | ||
| GenBank mRNA | JO415202.2 | - | 609 | 60 | ||
| JO429561.2 | - | 1089 | 60 | |||
More Information
Set Membership: |
|

UPB1
Genomic Map