Assay Details
Gene Symbol: PVR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062549.1 | XP_017918038.1 | 2 - 3 | 563 | 62 | |
XM_018062550.1 | XP_017918039.1 | 2 - 3 | 823 | 62 | ||
GenBank mRNA | JO540970.2 | - | 2 - 3 | 479 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
