Assay Details
| Gene Symbol: NTM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975868.2 | XP_013831322.1 | 5 - 6 | 980 | 60 | |
| XM_018043572.1 | XP_017899061.1 | 4 - 5 | 1493 | 60 | ||
| XM_018043573.1 | XP_017899062.1 | 4 - 5 | 1492 | 60 | ||
| XM_018043574.1 | XP_017899063.1 | 4 - 5 | 1489 | 60 | ||
| XM_018043575.1 | XP_017899064.1 | 5 - 6 | 911 | 60 | ||
| GenBank mRNA | JO646062.2 | - | 3 - 4 | 434 | 60 | |
More Information
Set Membership: |
|

NTM
Genomic Map