Assay Details
| Gene Symbol: CDK2AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043042.1 | XP_017898531.1 | 2 - 3 | 766 | 77 | |
| XM_018043045.1 | XP_017898534.1 | 2 - 3 | 283 | 77 | ||
| XM_018043046.1 | XP_017898535.1 | 2 - 3 | 473 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018043045.1, XM_018043046.1 transcripts.
Set Membership: |
|

CDK2AP2
Genomic Map