Assay Details
| Gene Symbol: CSNK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038614.1 | XP_017894103.1 | 6 - 7 | 885 | 122 | |
| GenBank mRNA | KA341983.1 | - | 1 - 2 | 528 | 122 | |
More Information
Set Membership: |
|

CSNK2B
Genomic Map