Assay Details
| Gene Symbol: HMGN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056431.1 | XP_017911920.1 | 5 - 6 | 475 | 86 | |
| GenBank mRNA | JO434200.2 | - | 5 - 6 | 376 | 86 | |
| JO437645.2 | - | 5 - 6 | 388 | 86 | ||
More Information
Set Membership: |
|

HMGN2
Genomic Map