Assay Details
Gene Symbol: ZNF354A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050558.1 | XP_017906047.1 | 3 - 4 | 1249 | 84 | |
XM_018050560.1 | XP_017906049.1 | 3 - 4 | 726 | 84 | ||
XM_018050561.1 | XP_017906050.1 | 3 - 4 | 1171 | 84 | ||
XM_018050562.1 | XP_017906051.1 | 3 - 4 | 412 | 84 | ||
GenBank mRNA | JT659514.1 | - | 2 - 3 | 805 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
