Assay Details
| Gene Symbol: SCAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695130.3 | XP_005695187.1 | 4 - 5 | 479 | 79 | |
| XM_005695131.3 | XP_005695188.1 | 4 - 5 | 446 | 79 | ||
| XM_018066235.1 | XP_017921724.1 | 5 - 6 | 609 | 79 | ||
| GenBank mRNA | JO428053.1 | - | 4 - 5 | 515 | 79 | |
More Information
Set Membership: |
|

SCAMP2
Genomic Map