Assay Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700374.3 | XP_005700431.1 | 14 - 15 | 2102 | 93 | |
| GenBank mRNA | JO580114.1 | - | 14 - 15 | 2158 | 93 | |
More Information
Set Membership: |
|

UTP14A
Genomic Map