Assay Details
| Gene Symbol: ATP6V1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685994.3 | XP_005686051.1 | 6 - 7 | 590 | 82 | |
| GenBank mRNA | JO646071.2 | - | 3 - 4 | 873 | 82 | |
More Information
Set Membership: |
|

ATP6V1D
Genomic Map