Assay Details
| Gene Symbol: INO80B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686329.3 | XP_005686386.2 | 1 - 2 | 146 | 114 | |
| GenBank mRNA | KA327107.1 | - | 1 - 2 | 116 | 114 | |
More Information
Set Membership: |
|

INO80B
Genomic Map