Assay Details
| Gene Symbol: RBIS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058172.1 | XP_017913661.1 | 3 - 4 | 489 | 89 | |
| GenBank mRNA | JO459922.2 | - | 1 - 2 | 126 | 89 | |
More Information
Set Membership: |
|

RBIS
Genomic Map