Assay Details
Gene Symbol: LOC102188265 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676440.2 | XP_005676497.2 | 2 | 317 | 91 |
Gene Symbol: LOC108635542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045644.1 | XP_017901133.1 | 3 | 336 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
