Assay Details
| Gene Symbol: HSPA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677146.3 | XP_005677203.2 | 1 | 1297 | 89 | |
| GenBank mRNA | JT645302.1 | - | 1 | 1343 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HSPA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677146.3 | XP_005677203.2 | 1 | 1297 | 89 | |
| GenBank mRNA | JT645302.1 | - | 1 | 1343 | 89 | |
Set Membership: |
|