Assay Details
| Gene Symbol: TMEM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039302.1 | XP_017894791.1 | 2 | 337 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM170B| Gene Symbol: TMEM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039302.1 | XP_017894791.1 | 2 | 337 | 105 | |
Set Membership: |
|