Assay Details
| Gene Symbol: ZBTB22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038581.1 | XP_017894070.1 | 2 | 1586 | 99 | |
| XM_018038582.1 | XP_017894071.1 | 2 | 1586 | 99 | ||
More Information
Set Membership: |
|

ZBTB22
Genomic Map