Assay Details
| Gene Symbol: MMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064076.1 | XP_017919565.1 | 7 | 2336 | 118 | |
| XM_018064081.1 | XP_017919570.1 | 6 | 2149 | 118 | ||
| XM_018064082.1 | XP_017919571.1 | 6 | 1996 | 118 | ||
More Information
Set Membership: |
|

MMD
Genomic Map