Assay Details
| Gene Symbol: MAGOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678393.3 | XP_005678450.1 | 5 | 619 | 110 | |
| GenBank mRNA | JO402054.2 | - | 1 | 65 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MAGOH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678393.3 | XP_005678450.1 | 5 | 619 | 110 | |
| GenBank mRNA | JO402054.2 | - | 1 | 65 | 110 | |
Set Membership: |
|