Assay Details
| Gene Symbol: GOLT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680790.3 | XP_005680847.2 | 5 | 1120 | 148 | |
| GenBank mRNA | JT652942.1 | - | 2 | 614 | 145 | |
More Information
Set Membership: |
|

GOLT1B
Genomic Map