Assay Details
| Gene Symbol: PDCD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO502308.2 | - | 6 | 1218 | 97 | |
| KA304584.1 | - | 1 | 91 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PDCD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO502308.2 | - | 6 | 1218 | 97 | |
| KA304584.1 | - | 1 | 91 | 97 | ||
Set Membership: |
|