Assay Details
| Gene Symbol: HINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050711.1 | XP_017906200.1 | 3 | 609 | 115 | |
| GenBank mRNA | JO445525.2 | - | 1 | 274 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HINT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050711.1 | XP_017906200.1 | 3 | 609 | 115 | |
| GenBank mRNA | JO445525.2 | - | 1 | 274 | 115 | |
Set Membership: |
|