Assay Details
| Gene Symbol: DNAJA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052362.1 | XP_017907851.1 | 9 | 1857 | 111 | |
| GenBank mRNA | KA339303.1 | - | 1 | 472 | 111 | |
More Information
Set Membership: |
|

DNAJA1
Genomic Map