Assay Details
| Gene Symbol: LOC102174895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005709559.3 | XP_005709616.3 | 2 | 474 | 98 | |
| XM_013962595.2 | XP_013818049.2 | 2 | 423 | 98 | ||
| GenBank mRNA | JO562565.2 | - | 3 | 345 | 98 | |
More Information
Set Membership: |
|

LOC102174895
Genomic Map