Assay Details
| Gene Symbol: CD151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042699.1 | XP_017898188.1 | 9 | 1619 | 63 | |
| XM_018042700.1 | XP_017898189.1 | 9 | 1569 | 63 | ||
| XM_018042702.1 | XP_017898191.1 | 9 | 1552 | 63 | ||
| GenBank mRNA | JO581836.1 | - | 2345 | 63 | ||
More Information
Set Membership: |
|

CD151
Genomic Map