Assay Details
| Gene Symbol: FILIP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674833.3 | XP_005674890.1 | 2 | 621 | 149 | |
| XM_013972898.2 | XP_013828352.1 | 5 | 1677 | 149 | ||
| XM_018058277.1 | XP_017913766.1 | 4 | 1197 | 149 | ||
| XM_018058386.1 | XP_017913875.1 | 2 | 732 | 149 | ||
| GenBank mRNA | JO365855.2 | - | 5 | 1597 | 149 | |
| JO365856.2 | - | 5 | 1597 | 149 | ||
More Information
Set Membership: |
|

FILIP1L
Genomic Map