Assay Details
| Gene Symbol: PGAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698285.3 | XP_005698342.2 | 4 | 1410 | 149 | |
| GenBank mRNA | JT752997.1 | - | 2 | 362 | 93 | |
| Gene Symbol: LOC102188713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047606.1 | XP_017903095.1 | 1 | 1293 | 134 | |
More Information
Set Membership: |
|

PGAM1
Genomic Map