Assay Details
| Gene Symbol: KCNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964275.2 | XP_013819729.2 | 1 | 1920 | 118 | |
| GenBank mRNA | JT667326.1 | - | 3 | 476 | 118 | |
More Information
Set Membership: |
|

KCNA5
Genomic Map