Assay Details
| Gene Symbol: MRPL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058137.1 | XP_017913626.1 | 7 | 677 | 104 | |
| XM_018058138.1 | XP_017913627.1 | 6 | 538 | 104 | ||
| XM_018058139.1 | XP_017913628.1 | 6 | 591 | 104 | ||
| GenBank mRNA | JO336764.2 | - | 1 | 174 | 104 | |
| JO618469.2 | - | 6 | 545 | 104 | ||
More Information
Set Membership: |
|

MRPL13
Genomic Map