Assay Details
| Gene Symbol: LOC102181107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681645.3 | XP_005681702.2 | 4 | 2168 | 123 | |
| XM_018049494.1 | XP_017904983.1 | 5 | 2289 | 123 | ||
| XM_018049495.1 | XP_017904984.1 | 4 | 2183 | 123 | ||
More Information
Set Membership: |
|

LOC102181107
Genomic Map