Assay Details
| Gene Symbol: FAM136A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055082.1 | XP_017910571.1 | 3 | 779 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM136A| Gene Symbol: FAM136A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055082.1 | XP_017910571.1 | 3 | 779 | 94 | |
Set Membership: |
|