Assay Details
| Gene Symbol: PHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044502.1 | XP_017899991.1 | 3 | 435 | 70 | |
| GenBank mRNA | JO514903.2 | - | 3 | 422 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044502.1 | XP_017899991.1 | 3 | 435 | 70 | |
| GenBank mRNA | JO514903.2 | - | 3 | 422 | 70 | |
Set Membership: |
|